ATC386058986 Atlantic Research Chemicals Ltd

5-Fluoro-6-methylpyridin-2-amine

  • MDL number MFCD09835298
  • Cas Number : 110919-71-6
  • Molecular Weight : 126.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://atlantic-chemicals.com/product/?code=AE002226
Availability details for
Quantity
  • Availability
  • Add to Cart