ATC386058996 Atlantic Research Chemicals Ltd

3-(3-Bromophenyl)-N-ethylacrylamide

  • MDL number MFCD00075351
  • Cas Number : 58473-74-8
  • Molecular Weight : 254.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://atlantic-chemicals.com/product/?code=XS001902
Availability details for
Quantity
  • Availability
  • Add to Cart