ATC386059032 Atlantic Research Chemicals Ltd

2-Pyrrolidin-1-ylisonicotinonitrile

  • MDL number MFCD08435908
  • Cas Number : 127680-87-9
  • Molecular Weight : 173.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://atlantic-chemicals.com/product/?code=XS002397
Availability details for
Quantity
  • Availability
  • Add to Cart