ATC386059062 Atlantic Research Chemicals Ltd

2-Methyl-4-(2-thienyl)-1,3-thiazole

  • MDL number MFCD01116539
  • Cas Number : 21036-67-9
  • Molecular Weight : 181.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://atlantic-chemicals.com/product/?code=XS00625
Availability details for
Quantity
  • Availability
  • Add to Cart