ATC386059071 Atlantic Research Chemicals Ltd

1,3-Dimethyl-1H-pyrazol-5-ol

  • MDL number MFCD00129673
  • Cas Number : 5203-77-0
  • Molecular Weight : 112.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://atlantic-chemicals.com/product/?code=XS00967
Availability details for
Quantity
  • Availability
  • Add to Cart