ATC386059082 Atlantic Research Chemicals Ltd

Tetrahydro-2H-pyran-4-yl 4-methylbenzenesulphonate

  • MDL number MFCD16620977
  • Cas Number : 97986-34-0
  • Molecular Weight : 256.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://atlantic-chemicals.com/product/?code=XS01424
Availability details for
Quantity
  • Availability
  • Add to Cart