ATC386059089 Atlantic Research Chemicals Ltd

1-(5-Nitro-2,3-dihydro-1H-indol-1-yl)ethan-1-one

  • MDL number MFCD00005811
  • Cas Number : 33632-27-8
  • Molecular Weight : 206.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://atlantic-chemicals.com/product/?code=XS01939
Availability details for
Quantity
  • Availability
  • Add to Cart