ATC386059095 Atlantic Research Chemicals Ltd

5-Hydroxy-2,1,3-benzothiadiazole

  • MDL number MFCD00457955
  • Cas Number : 768-10-5
  • Molecular Weight : 152.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://atlantic-chemicals.com/product/?code=XS02302
Availability details for
Quantity
  • Availability
  • Add to Cart