ATC386059102 Atlantic Research Chemicals Ltd

6-Methylquinoxaline

  • MDL number MFCD00041001
  • Cas Number : 6344-72-5
  • Molecular Weight : 144.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://atlantic-chemicals.com/product/?code=XS02807
Availability details for
Quantity
  • Availability
  • Add to Cart