ATC386059107 Atlantic Research Chemicals Ltd

1-Cyclobutylpiperidin-4-ol

  • MDL number MFCD11878581
  • Cas Number : 869224-62-4
  • Molecular Weight : 155.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://atlantic-chemicals.com/product/?code=XS037106
Availability details for
Quantity
  • Availability
  • Add to Cart