ATC386059157 Atlantic Research Chemicals Ltd

Glycidyl isobutyl ether

  • MDL number MFCD00055638
  • Cas Number : 3814-55-9
  • Molecular Weight : 130.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://atlantic-chemicals.com/product/?code=XS099170
Availability details for
Quantity
  • Availability
  • Add to Cart