ATC386059179 Atlantic Research Chemicals Ltd

1-(2'-Fluoro[1,1'-biphenyl]-4-yl)propan-1-ol

  • MDL number MFCD00662442
  • Cas Number : 64820-95-7
  • Molecular Weight : 230.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://atlantic-chemicals.com/product/?code=XS100229
Availability details for
Quantity
  • Availability
  • Add to Cart