ATC413215591 Atlantic Research Chemicals Ltd

1-(5-Amino-2-hydroxyphenyl)ethanone

  • MDL number MFCD00100397
  • Cas Number : 50-80-6
  • Molecular Weight : 151.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://atlantic-chemicals.com/product/?code=AE001251
Availability details for
Quantity
  • Availability
  • Add to Cart