ATC413215606 Atlantic Research Chemicals Ltd

4-Ethylpyridin-2-amine

  • MDL number MFCD00129025
  • Cas Number : 33252-32-3
  • Molecular Weight : 122.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://atlantic-chemicals.com/product/?code=AE002236
Availability details for
Quantity
  • Availability
  • Add to Cart