ATC413215653 Atlantic Research Chemicals Ltd

5-(2-Phenyleth-1-ynyl)thiophene-2-carbaldehyde

  • MDL number MFCD00126374
  • Cas Number : 17257-10-2
  • Molecular Weight : 212.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://atlantic-chemicals.com/product/?code=AH00317
Availability details for
Quantity
  • Availability
  • Add to Cart