ATC413215834 Atlantic Research Chemicals Ltd

3-(2,4-Dimethoxyphenyl)-2-propenoyl chloride

  • MDL number MFCD11940064
  • Cas Number : 931107-10-7
  • Molecular Weight : 226.66

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://atlantic-chemicals.com/product/?code=CC001136
Availability details for
Quantity
  • Availability
  • Add to Cart