ATC413215904 Atlantic Research Chemicals Ltd

1-Methylpiperidine-4-carbonyl chloride

  • MDL number MFCD13173735
  • Cas Number : 41776-24-3
  • Molecular Weight : 161.63

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://atlantic-chemicals.com/product/?code=CC03705
Availability details for
Quantity
  • Availability
  • Add to Cart