ATC413215927 Atlantic Research Chemicals Ltd

Ethyl 4-Amino-3,5-dichlorobenzoate

  • MDL number MFCD00016636
  • Cas Number : 74878-31-2
  • Molecular Weight : 234.08

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://atlantic-chemicals.com/product/?code=ES001224
Availability details for
Quantity
  • Availability
  • Add to Cart