ATC413215930 Atlantic Research Chemicals Ltd

Methyl 3-Anilinopropanoate

  • MDL number MFCD00089334
  • Cas Number : 21911-84-2
  • Molecular Weight : 179.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://atlantic-chemicals.com/product/?code=ES001227
Availability details for
Quantity
  • Availability
  • Add to Cart