ATC413215943 Atlantic Research Chemicals Ltd

Methyl 4-[4-(trifluoromethyl)pyridin-2-yl]benzoate

  • MDL number MFCD18384865
  • Molecular Weight : 281.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://atlantic-chemicals.com/product/?code=ES001240
Availability details for
Quantity
  • Availability
  • Add to Cart