ATC413215988 Atlantic Research Chemicals Ltd

Ethyl 2-Amino-5-iodothiazole-4-carboxylate

  • MDL number MFCD11975710
  • Cas Number : 860646-12-4
  • Molecular Weight : 298.099

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin United Kingdom
Product Link https://atlantic-chemicals.com/product/?code=ES00630
Availability details for
Quantity
  • Availability
  • Add to Cart