ATC413216021 Atlantic Research Chemicals Ltd

(2S,3R)-Ethyl 4-hydroxypyrrolidine-2-carboxylate hydrochloride

  • MDL number MFCD00040589
  • Cas Number : 33996-30-4
  • Molecular Weight : 195.65

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://atlantic-chemicals.com/product/?code=ES04523
Availability details for
Quantity
  • Availability
  • Add to Cart