ATC413216054 Atlantic Research Chemicals Ltd

Ethyl 4-(Benzyloxy)-3-oxobutanoate

  • MDL number MFCD19388798
  • Cas Number : 67354-34-1
  • Molecular Weight : 236.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://atlantic-chemicals.com/product/?code=ES10194
Availability details for
Quantity
  • Availability
  • Add to Cart