ATC413216123 Atlantic Research Chemicals Ltd

8-Bromo-3-methyl-1H-purine-2,6(3H,7H)-dione

  • MDL number MFCD00462528
  • Cas Number : 93703-24-3
  • Molecular Weight : 245.04

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://atlantic-chemicals.com/product/?code=HA100172
Availability details for
Quantity
  • Availability
  • Add to Cart