ATC413216208 Atlantic Research Chemicals Ltd

4-Bromo-3,5-dimethylbenzonitrile

  • MDL number MFCD07369910
  • Cas Number : 75344-77-3
  • Molecular Weight : 210.07

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://atlantic-chemicals.com/product/?code=XS001927
Availability details for
Quantity
  • Availability
  • Add to Cart