ATC413216253 Atlantic Research Chemicals Ltd

3-Bromocatechol

  • MDL number MFCD00869768
  • Cas Number : 14381-51-2
  • Molecular Weight : 189.01

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://atlantic-chemicals.com/product/?code=XS001972
Availability details for
Quantity
  • Availability
  • Add to Cart