ATC413216281 Atlantic Research Chemicals Ltd

2-(4-Bromophenoxy)-3-nitropyridine

  • MDL number MFCD09932563
  • Cas Number : 229620-55-7
  • Molecular Weight : 295.09

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://atlantic-chemicals.com/product/?code=XS002421
Availability details for
Quantity
  • Availability
  • Add to Cart