ATC413216302 Atlantic Research Chemicals Ltd

4-Bromopyridine-2-carboxamide

  • MDL number MFCD04113813
  • Cas Number : 62150-46-3
  • Molecular Weight : 201.023

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin United Kingdom
Product Link https://atlantic-chemicals.com/product/?code=XS002442
Availability details for
Quantity
  • Availability
  • Add to Cart