ATC429238175 Atlantic Research Chemicals Ltd

Methyl 4-[(4-Formylphenoxy)methyl]benzoate

  • MDL number MFCD01893188
  • Cas Number : 124663-30-5
  • Molecular Weight : 270.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://atlantic-chemicals.com/product/?code=AH001222
Availability details for
Quantity
  • Availability
  • Add to Cart