ATC429238195 Atlantic Research Chemicals Ltd

N-Methyl-N-({6-[(methylamino)methyl]pyridin-2-yl}methyl)amine

  • MDL number MFCD13249723
  • Cas Number : 134956-67-5
  • Molecular Weight : 165.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://atlantic-chemicals.com/product/?code=AM00262
Availability details for
Quantity
  • Availability
  • Add to Cart