ATC429238266 Atlantic Research Chemicals Ltd

2-Chloro-4-methoxynicotinic acid

  • MDL number MFCD18378504
  • Cas Number : 394729-98-7
  • Molecular Weight : 187.58

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://atlantic-chemicals.com/product/?code=CA002431
Availability details for
Quantity
  • Availability
  • Add to Cart