ATC429238275 Atlantic Research Chemicals Ltd

Pyrazine-2,5-dicarboxylic acid

  • MDL number MFCD01630902
  • Cas Number : 122-05-4
  • Molecular Weight : 168.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://atlantic-chemicals.com/product/?code=CA01119
Availability details for
Quantity
  • Availability
  • Add to Cart