ATC429238309 Atlantic Research Chemicals Ltd

(R)-Azetidine-2-carboxylic acid

  • MDL number MFCD00800731
  • Cas Number : 7729-30-8
  • Molecular Weight : 101.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://atlantic-chemicals.com/product/?code=CA099131
Availability details for
Quantity
  • Availability
  • Add to Cart