ATC429238545 Atlantic Research Chemicals Ltd

2,6-Dichlorophenylacetamide

  • MDL number MFCD00082745
  • Cas Number : 78433-88-2
  • Molecular Weight : 204.06

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://atlantic-chemicals.com/product/?code=XS0011012
Availability details for
Quantity
  • Availability
  • Add to Cart