ATC429238634 Atlantic Research Chemicals Ltd

4-(Trifluoromethyl)nicotinamide

  • MDL number MFCD00126658
  • Cas Number : 158062-71-6
  • Molecular Weight : 190.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://atlantic-chemicals.com/product/?code=XS002448
Availability details for
Quantity
  • Availability
  • Add to Cart