ATC429238651 Atlantic Research Chemicals Ltd

5-Aminonicotinamide

  • MDL number MFCD09038483
  • Cas Number : 60524-14-3
  • Molecular Weight : 137.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://atlantic-chemicals.com/product/?code=XS002465
Availability details for
Quantity
  • Availability
  • Add to Cart