ATC429238687 Atlantic Research Chemicals Ltd

2,3,5,6-Tetrafluoro-1,4-phenylenediamine

  • MDL number MFCD00039767
  • Cas Number : 1198-64-7
  • Molecular Weight : 180.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://atlantic-chemicals.com/product/?code=AE001268
Availability details for
Quantity
  • Availability
  • Add to Cart