ATC429238751 Atlantic Research Chemicals Ltd

5-Oxo-1-pyrrolidine-3-carboxamide

  • MDL number MFCD03613706
  • Cas Number : 14466-21-8
  • Molecular Weight : 128.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://atlantic-chemicals.com/product/?code=XS04528
Availability details for
Quantity
  • Availability
  • Add to Cart