ATC429238795 Atlantic Research Chemicals Ltd

Ethyl 2-Methoxyethanimidoate hydrochloride

  • MDL number MFCD25963422
  • Cas Number : 42945-65-3
  • Molecular Weight : 153.61

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://atlantic-chemicals.com/product/?code=XS101175
Availability details for
Quantity
  • Availability
  • Add to Cart