ATC456776421 Atlantic Research Chemicals Ltd

N'-(4-Aminophenyl)benzohydrazide

  • MDL number MFCD28976348
  • Cas Number : 63402-27-7
  • Molecular Weight : 227.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://atlantic-chemicals.com/product/?code=AE001287
Availability details for
Quantity
  • Availability
  • Add to Cart