ATC456776487 Atlantic Research Chemicals Ltd

Tert-Butyl 3-Aminoazepane-1-carboxylate

  • MDL number MFCD06738754
  • Cas Number : 609789-17-5
  • Molecular Weight : 214.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://atlantic-chemicals.com/product/?code=AE09944
Availability details for
Quantity
  • Availability
  • Add to Cart