ATC456776551 Atlantic Research Chemicals Ltd

3-Chloro-5-(trifluoromethoxy)phenylboronic acid

  • MDL number MFCD11856024
  • Cas Number : 1451393-40-0
  • Molecular Weight : 240.37

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://atlantic-chemicals.com/product/?code=BA00195
Availability details for
Quantity
  • Availability
  • Add to Cart