ATC456776573 Atlantic Research Chemicals Ltd

2-(2-Fluorobenzamido)acetic acid

  • MDL number MFCD00460020
  • Cas Number : 363-34-8
  • Molecular Weight : 197.164

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin United Kingdom
Product Link https://atlantic-chemicals.com/product/?code=CA001673
Availability details for
Quantity
  • Availability
  • Add to Cart