ATC456776620 Atlantic Research Chemicals Ltd

2-Ethoxycinnamic acid

  • MDL number MFCD00016839
  • Cas Number : 69038-81-9
  • Molecular Weight : 192.213

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin United Kingdom
Product Link https://atlantic-chemicals.com/product/?code=CA001720
Availability details for
Quantity
  • Availability
  • Add to Cart