ATC456777028 Atlantic Research Chemicals Ltd

Diethyl Furan-2,5-dicarboxylate

  • MDL number MFCD00449457
  • Cas Number : 53662-83-2
  • Molecular Weight : 212.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin United Kingdom
Product Link https://atlantic-chemicals.com/product/?code=ES00413
Availability details for
Quantity
  • Availability
  • Add to Cart