ATC456777042 Atlantic Research Chemicals Ltd

Ethyl 4-Cyclopropyl-1,3-thiazole-2-carboxylate

  • MDL number MFCD11656780
  • Cas Number : 439692-05-4
  • Molecular Weight : 197.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://atlantic-chemicals.com/product/?code=ES00637
Availability details for
Quantity
  • Availability
  • Add to Cart