ATC456777361 Atlantic Research Chemicals Ltd

1-(4-Chloromethylphenyl)-1H-pyrrole

  • MDL number MFCD20637779
  • Cas Number : 1187212-07-2
  • Molecular Weight : 191.66

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://atlantic-chemicals.com/product/?code=MH00151
Availability details for
Quantity
  • Availability
  • Add to Cart