ATC456777500 Atlantic Research Chemicals Ltd

6-(Trifluoromethyl)pyridine-2-carboxamide

  • MDL number MFCD02180820
  • Cas Number : 22245-84-7
  • Molecular Weight : 190.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://atlantic-chemicals.com/product/?code=XS002500
Availability details for
Quantity
  • Availability
  • Add to Cart