ATC456777600 Atlantic Research Chemicals Ltd

5-Thioxo-3,4-dihydro-1,2,4-triazin-3(2H)-one

  • MDL number MFCD00182966
  • Cas Number : 1627-37-8
  • Molecular Weight : 129.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://atlantic-chemicals.com/product/?code=XS05304
Availability details for
Quantity
  • Availability
  • Add to Cart