ATC491430501 Atlantic Research Chemicals Ltd

2-Phenyl-5-(trifluoromethyl)pyrazol-3-ol

  • MDL number MFCD03714773
  • Cas Number : 96145-98-1
  • Molecular Weight : 228.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://atlantic-chemicals.com/product/?code=XS00986
Availability details for
Quantity
  • Availability
  • Add to Cart